Assay Details
| Gene Symbol: top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001838.1 | NP_001001838.1 | 4 - 5 | 506 | 67 | |
| GenBank mRNA | AY429655.1 | - | 4 - 5 | 506 | 67 | |
| BC096851.1 | - | 4 - 5 | 506 | 67 | ||
More Information
Set Membership: |
|

top1mt
Genomic Map