Assay Details
| Gene Symbol: isl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002043.1 | NP_001002043.1 | 4 - 5 | 794 | 83 | |
| GenBank mRNA | AY423019.1 | - | 4 - 5 | 794 | 83 | |
More Information
Set Membership: |
|

isl1l
Genomic Map