Assay Details
| Gene Symbol: tmed4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002134.1 | NP_001002134.1 | 3 - 4 | 395 | 80 | |
| GenBank mRNA | BC071427.1 | - | 3 - 4 | 395 | 80 | |
More Information
Set Membership: |
|

tmed4
Genomic Map