Assay Details
Gene Symbol: dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002205.1 | NP_001002205.1 | 6 - 7 | 692 | 66 | |
GenBank mRNA | BC072703.1 | - | 6 - 7 | 692 | 66 | |
BC164751.1 | - | 5 - 6 | 682 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
