Assay Details
| Gene Symbol: rhbdf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002228.1 | NP_001002228.1 | 4 - 5 | 782 | 108 | |
| GenBank mRNA | BC074097.1 | - | 4 - 5 | 782 | 108 | |
More Information
Set Membership: |
|

rhbdf1a
Genomic Map