Assay Details
| Gene Symbol: rhbdf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002228.1 | NP_001002228.1 | 9 - 10 | 1632 | 93 | |
| XM_005164335.4 | XP_005164392.1 | 10 - 11 | 1940 | 93 | ||
| GenBank mRNA | BC074097.1 | - | 9 - 10 | 1632 | 93 | |
More Information
Set Membership: |
|

rhbdf1a
Genomic Map