Assay Details
Gene Symbol: bpnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002354.1 | NP_001002354.1 | 2 - 3 | 190 | 62 | |
XM_021468396.1 | XP_021324071.1 | 2 - 3 | 236 | 62 | ||
GenBank mRNA | BC075903.1 | - | 2 - 3 | 190 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
