Assay Details
| Gene Symbol: galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002373.1 | NP_001002373.1 | 2 - 3 | 413 | 62 | |
| XM_021480558.1 | XP_021336233.1 | 2 - 3 | 454 | 62 | ||
| GenBank mRNA | BC076039.1 | - | 2 - 3 | 413 | 62 | |
| BC153585.1 | - | 2 - 3 | 406 | 62 | ||
More Information
Set Membership: |
|

galm
Genomic Map