Assay Details
| Gene Symbol: nr4a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002406.1 | NP_001002406.1 | 3 - 4 | 1082 | 138 | |
| GenBank mRNA | AY542373.2 | - | 3 - 4 | 1082 | 138 | |
| BC076176.1 | - | 3 - 4 | 1082 | 138 | ||
More Information
Set Membership: |
|

nr4a2b
Genomic Map