Assay Details
| Gene Symbol: smpd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030219.2 | NP_001025390.2 | 1 - 2 | 66 | 73 | |
| XM_005170835.4 | XP_005170892.1 | 2 - 3 | 322 | 73 | ||
| GenBank mRNA | AB196165.1 | - | 1 - 2 | 46 | 73 | |
| BC163237.1 | - | 1 - 2 | 73 | 73 | ||
| EH590607.1 | - | 1 - 2 | 75 | 73 | ||
More Information
Set Membership: |
|

smpd2a
Genomic Map