Assay Details
| Gene Symbol: ada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033717.2 | NP_001028889.2 | 3 - 4 | 646 | 140 | |
| GenBank mRNA | AF384217.1 | - | 3 - 4 | 646 | 140 | |
| BC045839.1 | - | 3 - 4 | 621 | 140 | ||
More Information
Set Membership: |
|

ada2a
Genomic Map