Assay Details
| Gene Symbol: ada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033717.2 | NP_001028889.2 | 4 - 5 | 859 | 88 | |
| GenBank mRNA | AF384217.1 | - | 4 - 5 | 859 | 88 | |
More Information
Set Membership: |
|

ada2a
Genomic Map