Assay Details
Gene Symbol: txnl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037123.3 | NP_001032200.1 | 2 - 3 | 387 | 87 | |
NM_001290146.1 | NP_001277075.1 | 2 - 3 | 387 | 87 | ||
XM_021475835.1 | XP_021331510.1 | 2 - 3 | 391 | 87 | ||
GenBank mRNA | BC107846.1 | - | 2 - 3 | 341 | 87 | |
EH437899.1 | - | 2 - 3 | 384 | 87 | ||
EH498384.1 | - | 2 - 3 | 398 | 87 | ||
EH591445.1 | - | 2 - 3 | 418 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
