Assay Details
| Gene Symbol: nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037704.2 | NP_001032793.2 | 2 - 3 | 505 | 115 | |
| GenBank mRNA | AB178827.1 | - | 2 - 3 | 196 | 115 | |
| BC124089.1 | - | 2 - 3 | 257 | 115 | ||
| BC162854.1 | - | 2 - 3 | 247 | 115 | ||
| BC162880.1 | - | 2 - 3 | 245 | 115 | ||
| EH553755.1 | - | 2 - 3 | 351 | 115 | ||
| KM655646.1 | - | 2 - 3 | 196 | 115 | ||
More Information
Set Membership: |
|

nenf
Genomic Map