Assay Details
| Gene Symbol: inpp4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039973.2 | NP_001035062.2 | 9 - 10 | 1022 | 83 | |
| XM_017357717.2 | XP_017213206.1 | 9 - 10 | 1043 | 83 | ||
| GenBank mRNA | BC114330.1 | - | 8 - 9 | 923 | 83 | |
More Information
Set Membership: |
|

inpp4aa
Genomic Map