Assay Details
| Gene Symbol: fgf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040095.1 | NP_001035184.1 | 1 - 2 | 320 | 87 | |
| GenBank mRNA | AB254028.1 | - | 1 - 2 | 320 | 87 | |
| BC162959.1 | - | 1 - 2 | 414 | 87 | ||
| BC162995.1 | - | 1 - 2 | 414 | 87 | ||
More Information
Set Membership: |
|

fgf22
Genomic Map