Assay Details
Gene Symbol: top1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044324.2 | NP_001037789.1 | 11 - 12 | 1074 | 68 | |
GenBank mRNA | BC071418.1 | - | 11 - 12 | 1073 | 68 | |
BC117588.1 | - | 11 - 12 | 1074 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
