Assay Details
Gene Symbol: top1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044324.2 | NP_001037789.1 | 12 - 13 | 1263 | 67 | |
GenBank mRNA | BC071418.1 | - | 12 - 13 | 1262 | 67 | |
BC117588.1 | - | 12 - 13 | 1263 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
