Assay Details
Gene Symbol: top1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044324.2 | NP_001037789.1 | 17 - 18 | 1919 | 79 | |
GenBank mRNA | BC117588.1 | - | 17 - 18 | 1919 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
