Assay Details
Gene Symbol: top1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044324.2 | NP_001037789.1 | 3 - 4 | 272 | 150 | |
GenBank mRNA | BC071418.1 | - | 3 - 4 | 271 | 150 | |
BC095049.1 | - | 3 - 4 | 231 | 150 | ||
BC117588.1 | - | 3 - 4 | 272 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
