Assay Details
| Gene Symbol: ugt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044325.2 | NP_001037790.2 | 3 - 4 | 1119 | 146 | |
| GenBank mRNA | BC093324.1 | - | 3 - 4 | 1112 | 146 | |
| BC117632.1 | - | 1119 | 146 | |||
More Information
Set Membership: |
|

ugt8
Genomic Map