Assay Details
| Gene Symbol: fabp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044712.1 | NP_001038177.1 | 2 - 3 | 125 | 77 | |
| GenBank mRNA | BC114269.1 | - | 2 - 3 | 108 | 77 | |
| BC163809.1 | - | 2 - 3 | 163 | 77 | ||
| BC163810.1 | - | 2 - 3 | 149 | 77 | ||
| DQ062095.1 | - | 2 - 3 | 125 | 77 | ||
More Information
Set Membership: |
|

fabp1a
Genomic Map