Assay Details
| Gene Symbol: fabp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044712.1 | NP_001038177.1 | 3 - 4 | 301 | 70 | |
| GenBank mRNA | BC163809.1 | - | 3 - 4 | 339 | 70 | |
| BC163810.1 | - | 3 - 4 | 325 | 70 | ||
| DQ062095.1 | - | 3 - 4 | 301 | 70 | ||
More Information
Set Membership: |
|

fabp1a
Genomic Map