Assay Details
| Gene Symbol: nudt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044816.1 | NP_001038281.1 | 5 - 6 | 639 | 66 | |
| XM_009306499.2 | XP_009304774.1 | 7 - 8 | 1444 | 66 | ||
| XM_009306500.2 | XP_009304775.1 | 6 - 7 | 1319 | 66 | ||
More Information
Set Membership: |
|

nudt13
Genomic Map