Assay Details
Gene Symbol: grid2ipb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044903.1 | NP_001038368.1 | 2 - 3 | 363 | 67 | |
XM_009306633.3 | XP_009304908.1 | 6 - 7 | 1362 | 67 | ||
XM_017358463.2 | XP_017213952.2 | 6 - 7 | 1362 | 67 | ||
XM_021480122.1 | XP_021335797.1 | 6 - 7 | 1493 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
