Assay Details
| Gene Symbol: ccnd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045068.1 | NP_001038533.1 | 3 - 4 | 568 | 65 | |
| XM_005164521.4 | XP_005164578.1 | 4 - 5 | 842 | 65 | ||
| GenBank mRNA | FJ648074.1 | - | 3 - 4 | 631 | 65 | |
More Information
Set Membership: |
|

ccnd2b
Genomic Map