Assay Details
Gene Symbol: dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045276.2 | NP_001038741.2 | 9 - 10 | 1348 | 68 | |
XM_005158227.4 | XP_005158284.1 | 9 - 10 | 1396 | 68 | ||
XM_009292539.3 | XP_009290814.1 | 9 - 10 | 1403 | 68 | ||
GenBank mRNA | BC116512.1 | - | 9 - 10 | 1303 | 68 | |
BC155223.1 | - | 9 - 10 | 1349 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
