Assay Details
Gene Symbol: snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045374.2 | NP_001038839.2 | 2 - 3 | 367 | 73 | |
GenBank mRNA | BC122395.1 | - | 2 - 3 | 336 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
