Assay Details
| Gene Symbol: aimp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045851.1 | NP_001039316.1 | 5 - 6 | 723 | 62 | |
| GenBank mRNA | BC122449.1 | - | 5 - 6 | 723 | 62 | |
| BC164340.1 | - | 4 - 5 | 650 | 62 | ||
More Information
Set Membership: |
|

aimp1a
Genomic Map