Assay Details
| Gene Symbol: ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076555.1 | NP_001070023.1 | 1 - 2 | 188 | 64 | |
| GenBank mRNA | BC124536.1 | - | 1 - 2 | 188 | 64 | |
More Information
Set Membership: |
|

ifrd1
Genomic Map