Assay Details
| Gene Symbol: fam102ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076613.1 | NP_001070081.1 | 6 - 7 | 717 | 75 | |
| XM_009301550.3 | XP_009299825.1 | 6 - 7 | 1066 | 75 | ||
| XM_009301551.3 | XP_009299826.1 | 6 - 7 | 1067 | 75 | ||
| XM_009301552.3 | XP_009299827.1 | 6 - 7 | 1063 | 75 | ||
| GenBank mRNA | BC124191.1 | - | 6 - 7 | 717 | 75 | |
| BC164783.1 | - | 4 - 5 | 445 | 75 | ||
More Information
Set Membership: |
|

fam102ab
Genomic Map