Assay Details
| Gene Symbol: rps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076732.2 | NP_001070200.1 | 4 - 5 | 279 | 132 | |
| GenBank mRNA | BC122271.1 | - | 4 - 5 | 281 | 132 | |
| BC164282.1 | - | 3 - 4 | 281 | 132 | ||
More Information
Set Membership: |
|

rps16
Genomic Map