Assay Details
| Gene Symbol: lrrc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076768.1 | NP_001070236.1 | 2 - 3 | 477 | 85 | |
| XM_009291957.3 | XP_009290232.1 | 2 - 3 | 492 | 85 | ||
| GenBank mRNA | BC091531.1 | - | 2 - 3 | 464 | 85 | |
| BC124442.1 | - | 2 - 3 | 477 | 85 | ||
More Information
Set Membership: |
|

lrrc51
Genomic Map