Assay Details
| Gene Symbol: kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077151.1 | NP_001070619.1 | 2 - 3 | 442 | 55 | |
| GenBank mRNA | BC122284.1 | - | 2 - 3 | 442 | 55 | |
| BC157374.1 | - | 2 - 3 | 410 | 55 | ||
More Information
Set Membership: |
|

kctd13
Genomic Map