Assay Details
| Gene Symbol: minpp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077557.1 | NP_001071025.1 | 3 - 4 | 987 | 78 | |
| XM_005158515.4 | XP_005158572.1 | 3 - 4 | 1128 | 78 | ||
| GenBank mRNA | BC124216.1 | - | 3 - 4 | 987 | 78 | |
| BC164680.1 | - | 3 - 4 | 924 | 78 | ||
More Information
Set Membership: |
|

minpp1a
Genomic Map