Assay Details
Gene Symbol: minpp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077557.1 | NP_001071025.1 | 4 - 5 | 1117 | 70 | |
XM_005158515.4 | XP_005158572.1 | 4 - 5 | 1258 | 70 | ||
GenBank mRNA | BC124216.1 | - | 4 - 5 | 1117 | 70 | |
BC164680.1 | - | 4 - 5 | 1054 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
