Assay Details
| Gene Symbol: smpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077615.1 | NP_001071083.1 | 4 - 5 | 1398 | 62 | |
| XM_005158452.4 | XP_005158509.1 | 4 - 5 | 1625 | 62 | ||
| XM_005158453.4 | XP_005158510.1 | 4 - 5 | 1637 | 62 | ||
| GenBank mRNA | AB361066.1 | - | 3 - 4 | 1229 | 62 | |
| BC124788.1 | - | 4 - 5 | 1398 | 62 | ||
| BC155730.1 | - | 4 - 5 | 1410 | 62 | ||
| BC165400.1 | - | 3 - 4 | 1263 | 62 | ||
More Information
Set Membership: |
|

smpd5
Genomic Map