Assay Details
| Gene Symbol: hmgcra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079977.2 | NP_001073446.2 | 11 - 12 | 1574 | 97 | |
| XM_005165515.4 | XP_005165572.1 | 11 - 12 | 1549 | 97 | ||
| GenBank mRNA | BC117622.1 | - | 11 - 12 | 1578 | 97 | |
| BC155135.1 | - | 11 - 12 | 1549 | 97 | ||
More Information
Set Membership: |
|

hmgcra
Genomic Map