Assay Details
| Gene Symbol: adnpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080015.2 | NP_001073484.2 | 3 - 4 | 585 | 75 | |
| GenBank mRNA | BC128662.1 | - | 3 - 4 | 563 | 75 | |
| EB936572.1 | - | 3 - 4 | 356 | 75 | ||
More Information
Set Membership: |
|

adnpa
Genomic Map