Assay Details
| Gene Symbol: kdm2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081692.2 | NP_001075161.2 | 16 - 17 | 2209 | 85 | |
| XM_005160248.4 | XP_005160305.1 | 16 - 17 | 2167 | 85 | ||
| XM_009293357.3 | XP_009291632.1 | 16 - 17 | 2795 | 85 | ||
| XM_009293365.3 | XP_009291640.1 | 16 - 17 | 2096 | 85 | ||
| GenBank mRNA | BC128874.1 | - | 7 - 8 | 1005 | 85 | |
More Information
Set Membership: |
|

kdm2aa
Genomic Map