Assay Details
Gene Symbol: rdh10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080583.1 | NP_001074052.1 | 2 - 3 | 575 | 106 | |
GenBank mRNA | BC129239.1 | - | 2 - 3 | 575 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
