Assay Details
| Gene Symbol: csrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080614.1 | NP_001074083.1 | 5 - 6 | 562 | 87 | |
| GenBank mRNA | BC129417.1 | - | 5 - 6 | 562 | 87 | |
| BC163943.1 | - | 4 - 5 | 542 | 87 | ||
More Information
Set Membership: |
|

csrp1b
Genomic Map