Assay Details
Gene Symbol: sacm1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080624.2 | NP_001074093.2 | 15 - 16 | 1337 | 100 | |
GenBank mRNA | BC129344.1 | - | 15 - 16 | 1337 | 100 | |
BC165691.1 | - | 15 - 16 | 1331 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
