Assay Details
| Gene Symbol: slc26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080667.1 | NP_001074136.1 | 1 - 2 | 629 | 72 | |
| XM_005161206.4 | XP_005161263.1 | 3 - 4 | 958 | 72 | ||
| XM_009295333.3 | XP_009293608.1 | 3 - 4 | 955 | 72 | ||
| GenBank mRNA | BC129231.1 | - | 1 - 2 | 629 | 72 | |
More Information
Set Membership: |
|

slc26a1
Genomic Map