Assay Details
| Gene Symbol: slc32a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080701.1 | NP_001074170.1 | 1 - 2 | 805 | 65 | |
| GenBank mRNA | BC129202.1 | - | 1 - 2 | 805 | 65 | |
More Information
Set Membership: |
|

slc32a1
Genomic Map