Assay Details
Gene Symbol: kdm2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081692.2 | NP_001075161.2 | 7 - 8 | 891 | 95 | |
XM_005160248.4 | XP_005160305.1 | 7 - 8 | 849 | 95 | ||
XM_009293357.3 | XP_009291632.1 | 7 - 8 | 1477 | 95 | ||
XM_009293362.3 | XP_009291637.1 | 7 - 8 | 848 | 95 | ||
XM_009293365.3 | XP_009291640.1 | 7 - 8 | 778 | 95 | ||
GenBank mRNA | BC129321.1 | - | 7 - 8 | 893 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
