Assay Details
| Gene Symbol: taco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082873.2 | NP_001076342.2 | 2 - 3 | 624 | 73 | |
| GenBank mRNA | BC133903.1 | - | 2 - 3 | 394 | 73 | |
| EH583816.1 | - | 2 - 3 | 619 | 73 | ||
| EH601240.1 | - | 3 - 4 | 700 | 73 | ||
More Information
Set Membership: |
|

taco1
Genomic Map