Assay Details
| Gene Symbol: mfsd6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083056.1 | NP_001076525.1 | 4 - 5 | 1825 | 63 | |
| XM_005161679.4 | XP_005161736.1 | 5 - 6 | 2794 | 63 | ||
| XM_021469281.1 | XP_021324956.1 | 4 - 5 | 1825 | 63 | ||
More Information
Set Membership: |
|

mfsd6a
Genomic Map