Assay Details
| Gene Symbol: lrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083068.1 | NP_001076537.1 | 4 - 5 | 1088 | 58 | |
| XM_005159560.3 | XP_005159617.1 | 4 - 5 | 1333 | 58 | ||
| XM_009294101.1 | XP_009292376.1 | 3 - 4 | 399 | 58 | ||
| GenBank mRNA | BC161621.1 | - | 4 - 5 | 1325 | 58 | |
More Information
Set Membership: |
|

lrp12
Genomic Map