Assay Details
| Gene Symbol: ghra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083578.1 | NP_001077047.1 | 7 - 8 | 1048 | 137 | |
| GenBank mRNA | BC134903.1 | - | 7 - 8 | 1048 | 137 | |
| GQ890150.1 | - | 1 - 2 | 92 | 137 | ||
| GQ890151.1 | - | 1 - 2 | 92 | 137 | ||
More Information
Set Membership: |
|

ghra
Genomic Map